accessibility menu, dialog, popup

UserName

KH3COSS

CAS RN 2720-67-4

IUPAC Name: potassium (propoxymethanethioyl)sulfanide
Synonyms: potassium (propoxymethanethioyl)sulfanide
Molecular Weight (g/mol): 174.32
Molecular Formula: C4H7KOS2
InChi Key: NZUFLKMNMAHESJ-UHFFFAOYSA-M
SMILES: [K+].CCCOC([S-])=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results