missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Propionate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 112.17 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Potassium Propionate |
Safety and Handling
| EINECSNumber | (2)-4132 |
| TSCA | No |