accessibility menu, dialog, popup

UserName

KH3COO

CAS RN 327-62-8

IUPAC Name: potassium propanoate
Synonyms: potassium propionate
Molecular Weight (g/mol): 112.17
Molecular Formula: C3H5KO2
InChi Key: BWILYWWHXDGKQA-UHFFFAOYSA-M
SMILES: [K+].CCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results