missing translation for 'onlineSavingsMsg'
Learn More

Potassium phthalate, Monobasic, Certified Reference Material, For titrimetry, Certified by BAM, 99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015732
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015732 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015732 Supplier Merck Emd Millipore Supplier No. 6035750G

Potassium phthalate monobasic can find applications in acid and base titrimetric analysis as an acidimetric standard and pH standard.

Specifications

CAS 877-24-7
Physical Form Neat
Linear Formula HOOCC6H4COOK
MDL Number MFCD00013070
Melting Point 295°C to 300°C (decomposition)
Synonym KHP; Phthalic acid monopotassium salt; Potassium biphthalate; Potassium hydrogen phthalate
Quantity 50 g
Formula Weight 204.22
Percent Purity ≥99.5%
Grade Certified Reference Material