missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Tribasic, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$34587.44 - $34587.44
Chemical Identifiers
| CAS | 7778-53-2 |
|---|---|
| Molecular Formula | K3O4P |
| Molecular Weight (g/mol) | 212.265 |
| InChI Key | LWIHDJKSTIGBAC-UHFFFAOYSA-K |
| Synonym | potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 |
| PubChem CID | 62657 |
| IUPAC Name | tripotassium;phosphate |
| SMILES | [O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02003803
|
Avantor J.T.Baker
325905 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7778-53-2 | |
| 212.265 | |
| potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
| tripotassium;phosphate |
| K3O4P | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| 62657 | |
| [O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
Specifications
| 7778-53-2 | |
| 2.5 kg | |
| potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
| [O-]P(=O)([O-])[O-].[K+].[K+].[K+] | |
| 212.265 | |
| BAKER ANALYZED™ Reagent | |
| Potassium Phosphate, Tribasic, n-Hydrate |
| Powder | |
| K3O4P | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| tripotassium;phosphate | |
| 62657 | |
| Poly Bottle |