missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Tribasic, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Phosphate, Tribasic, n-Hydrate |