missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, ULTREX™ Ultrapure Reagent, J.T. Baker™

Catalog No. p-5390855
Change view
Click to view available options
Quantity:
1 kg
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02003812 100 g Wide Mouth Amber Glass Bottle
02003814 1 kg Wide Mouth Amber Glass Bottle
2 options

Catalog No. 02003812

Supplier: Avantor 492104

Only null left

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Quantity 100 g
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09
Grade ULTREX™ Ultrapure Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Potassium Phosphate, Monobasic