missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, ULTREX™ Ultrapure Reagent, J.T. Baker™
Specifications
Specifications
| Quantity | 1 kg |
| IUPAC Name | potassium dihydrogen phosphate |
| Formula Weight | 136.09 |
| Grade | ULTREX™ Ultrapure Reagent |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.34kg |
| Chemical Name or Material | Potassium Phosphate, Monobasic |