missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, for HPLC, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 50 g |
| Linear Formula | KH2PO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 136.09g/mol |
| Percent Purity | ≥99.5% |
| Density | 2.338g/mL at 25°C |
| Chemical Name or Material | Potassium phosphate monobasic |
Safety and Handling
| RTECSNumber | TC6615500 |