missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, Crystal, Multi-Compendial, N.F., J.T. Baker™

Catalog No. 02003801
Change view
Click to view available options
Quantity:
2.5 kg
500 g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02003801 500 g Wide Mouth Amber Glass Bottle
02003799 2.5 kg Poly Bottle
02003798 2.5 kg Wide Mouth Amber Glass Bottle
3 options

Catalog No. 02003801

Supplier: Avantor 324801

Only null left

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Grade Multi-Compendial/NF
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Potassium Phosphate, Monobasic