missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, Crystal, Multi-Compendial, N.F., J.T. Baker™

Catalog No. 02003799
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003799 2.5 kg Poly Bottle
02003801 500 g Wide Mouth Amber Glass Bottle
02003798 2.5 kg Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003799 Supplier Avantor Supplier No. 324815

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-77-0 | Formula: H2KO4P | Weight: 136.08 g/mol

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystals
Quantity 2.5 kg
Packaging Poly Bottle
Specific Gravity 1L = 2.34kg
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09
Grade Multi-Compendial/NF
Chemical Name or Material Potassium Phosphate, Monobasic