missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
$89.47 - $950.83
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
| PubChem CID | 516951 |
| ChEBI | CHEBI:63036 |
| IUPAC Name | potassium dihydrogen phosphate |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN1914301
|
MP Biomedicals Inc
021914301 |
100 g |
Each for $89.47
|
|
|||||
|
ICN19143001
|
MP Biomedicals Inc
0219143001 |
1 kg |
Each for $368.49
|
|
|||||
|
ICN19143005
|
MP Biomedicals Inc
0219143005 |
5 kg |
Each for $950.83
|
|
|||||
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| 516951 | |
| potassium dihydrogen phosphate |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| CHEBI:63036 | |
| [K+].OP(O)([O-])=O |
Specifications
| 7778-77-0 | |
| H2KO4P | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| CHEBI:63036 |
| 100 g | |
| MFCD00011401 MFCD00147253 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 516951 |