missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic Anhydrous, MP Biomedicals

Catalog No. ICN1914301
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1914301 100 g
ICN19143001 1 kg
ICN19143005 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN1914301 Supplier MP Biomedicals Inc Supplier No. 02191430.1
Only null left

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Quantity 100 g