missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals

Supplier:  MP Biomedicals Inc 0219472790

 View more versions of this product

Catalog No. ICN19472790


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Phosphate Monobasic
7778-77-0
500 g
H2KO4P
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
Anhydrous
Crystalline
99+%
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
516951
136.09

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium dihydrogen phosphate
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
[K+].OP(O)([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions