missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, monobasic, 99%, for analysis

Catalog No. AC447670010
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447670010 1 kg
AC447670050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447670010 Supplier Thermo Scientific Chemicals Supplier No. 447670010

missing translation for 'validMainframeMsg'

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 99%
Linear Formula KH2PO4
Merck Index 15,778
Solubility Information Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 136.09
Percent Purity 99%
Grade Analytical
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less
EINECSNumber 231-913-4

RUO – Research Use Only