missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Trihydrate, (White, crystalline powder/Reagent Grade), MP Biomedicals

Catalog No. ICN19143201
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19143201 1 kg
ICN19143205 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19143201 Supplier MP Biomedicals Inc Supplier No. 0219143201

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Formula Weight 228.2
Grade Reagent
Chemical Name or Material Potassium Phosphate Dibasic Trihydrate
Recommended Storage Room Temperature