missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic, Trihydrate, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18605666
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605666 500 g Poly Bottle
18605665 125 g Poly Bottle
18605668 2.5 kg Poly Pail
18605667 6 x 500 g Poly Bottle
18605669 4 x 2.5 kg Poly Pail
18605670 12 kg Poly Pail
18605671 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18605666 Supplier Spectrum Chemical Mfg Cor Supplier No. P1385500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1385, 16788-57-1, K2HPO4·3H2O

Spectrum™ Chemical Potassium Phosphate Dibasic, Trihydrate, Reagent is a soluble salt that is commonly used as a buffering agent. As a Reagent Grade, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

CAS Max % 1
pH 9.0 to 9.4
Quantity 500 g
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle