missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade, Calbiochem™,

$423.00 - $423.00

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5295671KG
View Documents
MilliporeSigma™
5295671KG
1 kg N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16788-57-1
228.22
XQGPKZUNMMFTAL-UHFFFAOYSA-L
16217523
O.O.O.[K+].[K+].OP([O-])([O-])=O
H7K2O7P
MFCD00149926
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
dipotassium trihydrate hydrogen phosphate

Specifications

16788-57-1
≥99%
MFCD00149926
Easily soluble in cold water
O.O.O.[K+].[K+].OP([O-])([O-])=O
228.22
228.2
by titration
1 kg
H7K2O7P
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
XQGPKZUNMMFTAL-UHFFFAOYSA-L
dipotassium trihydrate hydrogen phosphate
16217523
Potassium Phosphate, Dibasic, Trihydrate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).

SDS
missing translation for 'documents'

missing translation for 'documents'