missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, dibasic trihydrate, 99+%, for analysis

Catalog No. p-43449
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205935000 500 g Plastic bottle
AC205930025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC205935000 Supplier Thermo Scientific Chemicals Supplier No. 205935000
Only null left

CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Color White
pH 8.5 to 9.6 (5% soln.)
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula K2HPO4·3H2O
Merck Index 10, 7541
Solubility Information Solubility in water: freely soluble
IUPAC Name dipotassium;hydrogen phosphate;trihydrate
Formula Weight 228.23
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic trihydrate, 99+%
Show More Show Less

RUO – Research Use Only