missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium phosphate, 0.5M buffer soln., pH 7.5

CAS: 7778-77-0 | H2KO4P | 136.084 g/mol

$174.41 - $270.10

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.084
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61553AK
View Documents
Thermo Scientific Chemicals
J61553AK
250 mL
Each for $174.41
Only null left
 
AAJ61553AP
View Documents
Thermo Scientific Chemicals
J61553AP
500 mL
Each for $270.10
Only null left
 
Description

Description

Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble in ethanol, alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Specifications

Chemical Identifiers

7778-77-0
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
[K+].OP(O)([O-])=O
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate

Specifications

7778-77-0
7.5
250 mL
MFCD00011401 MFCD00147253
[K+].OP(O)([O-])=O
136.084
0.5M buffer soln.
Liquid
H2KO4P
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only