missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Potassium phosphate, 0.2M buffer soln., pH 7.5, low endotoxin

$123.22 - $123.22

Specifications

CAS 7778-77-0
Color Colorless
Concentration 0.2M buffer soln.
pH 7.5
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67297AK
View Documents
Thermo Scientific™
J67297AK
250 mL
Each for $123.22
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

7778-77-0
0.2M buffer soln.
Liquid
MFCD00011401 MFCD00147253
[K+].OP(O)([O-])=O
136.084
Colorless
7.5
H2KO4P
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'