missing translation for 'onlineSavingsMsg'
Learn More

Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog No. p-7031084
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3203003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3203003 Supplier Thermo Scientific Chemicals Supplier No. H3203003
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1027642-30-3
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD10566516
InChI Key OVYSNXCRBZPJIG-UHFFFAOYSA-N
Synonym potassium phenoxy-methyltrifluoroborate, potassium trifluoro phenoxymethyl boranuide, pubchem11600, potassium phenoxymethyltrifluoroborate, potassium trifluoro phenoxymethyl borate, potassium trifluoro phenoxymethyl borate 1-, potassium phenoxymethyltrifluoroborate, contains kbr
PubChem CID 45479874
IUPAC Name potassium;trifluoro(phenoxymethyl)boranuide
SMILES [B-](COC1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Quantity 1 g
Formula Weight 214.04
Percent Purity 95%
Chemical Name or Material Potassium phenoxymethyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only