missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), Reagents
Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C12886001KG
Specifications
| Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate) | |
| Colorless/White | |
| Solid | |
| K2O8S2 | |
| USHAGKDGDHPEEY-UHFFFAOYSA-L | |
| dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| ACS | |
| 2.477 |
| >170°C | |
| 7727-21-1 | |
| 1 kg | |
| Potassium Peroxydisulfate, Potassium Peroxodisulphate | |
| [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 270.31 | |
| Poly Bottle |
Chemical Identifiers
| 7727-21-1 | |
| 270.31 | |
| Potassium Peroxydisulfate, Potassium Peroxodisulphate | |
| [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| K2O8S2 | |
| USHAGKDGDHPEEY-UHFFFAOYSA-L | |
| dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide |