missing translation for 'onlineSavingsMsg'
Learn More

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), Reagents

Catalog No. p-200020882
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337807 10 kg
01337804 500 g
01337806 125 g
01337805 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337807 Supplier Labchem Inc Supplier No. C128860010KG
Only null left

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.31
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym Potassium Peroxydisulfate, Potassium Peroxodisulphate
IUPAC Name dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point >170°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 2.477
Chemical Name or Material Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate)