missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Periodate, ACS Reagent (Potassium Metaperiodate), Reagents
Potassium Periodate, ACS Reagent (Potassium Metaperiodate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C128820025KG1
Specifications
| 300°C (loses oxygen) | |
| White | |
| Solid | |
| IKO4 | |
| FJVZDOGVDJCCCR-UHFFFAOYSA-M | |
| potassium periodate | |
| ACS | |
| 3.618 |
| Potassium Periodate, ACS Reagent (Potassium Metaperiodate) | |
| 7790-21-8 | |
| 2.5 kg | |
| Potassium Metaperiodate | |
| [K+].[O-][I](=O)(=O)=O | |
| 230.00 | |
| Poly Bottle |
Chemical Identifiers
| 7790-21-8 | |
| 230.00 | |
| Potassium Metaperiodate | |
| [K+].[O-][I](=O)(=O)=O |
| IKO4 | |
| FJVZDOGVDJCCCR-UHFFFAOYSA-M | |
| potassium periodate |