missing translation for 'onlineSavingsMsg'
Learn More

Potassium Periodate, ACS Reagent (Potassium Metaperiodate), Reagents

Catalog No. p-200021056
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
25 kg
Catalog No. Quantity
01337816 10 kg
01337815 2.5 kg
01337814 25 kg
01337813 500 g
4 options

Catalog No. 01337816

Supplier: Labchem Inc C128820010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Periodate, ACS Reagent (Potassium Metaperiodate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 230.00
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym Potassium Metaperiodate
IUPAC Name potassium periodate
SMILES [K+].[O-][I](=O)(=O)=O

Specifications

Boiling Point 300°C (loses oxygen)
Color White
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 3.618
Chemical Name or Material Potassium Periodate, ACS Reagent (Potassium Metaperiodate)