missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium p-Toluenethiosulfonate 98.0+%, TCI America™
$90.48 - $236.80
Chemical Identifiers
| CAS | 28519-50-8 |
|---|---|
| Molecular Formula | C7H7KO2S2 |
| Molecular Weight (g/mol) | 226.349 |
| MDL Number | MFCD00003547 |
| InChI Key | RUDNWZFWWJFUSF-UHFFFAOYSA-M |
| Synonym | p-Toluenethiosulfonic Acid Potassium Salt |
| PubChem CID | 23662126 |
| IUPAC Name | potassium;(4-methylphenyl)-oxido-oxo-sulfanylidene-$l^{6}-sulfane |
| SMILES | CC1=CC=C(C=C1)S(=O)(=S)[O-].[K+] |
Chemical Identifiers
| 28519-50-8 | |
| 226.349 | |
| RUDNWZFWWJFUSF-UHFFFAOYSA-M | |
| 23662126 | |
| CC1=CC=C(C=C1)S(=O)(=S)[O-].[K+] |
| C7H7KO2S2 | |
| MFCD00003547 | |
| p-Toluenethiosulfonic Acid Potassium Salt | |
| potassium;(4-methylphenyl)-oxido-oxo-sulfanylidene-$l^{6}-sulfane |
Specifications
| 28519-50-8 | |
| White | |
| MFCD00003547 | |
| p-Toluenethiosulfonic Acid Potassium Salt | |
| CC1=CC=C(C=C1)S(=O)(=S)[O-].[K+] | |
| 226.349 | |
| 226.35 | |
| Crystalline Powder |
| 229°C | |
| C7H7KO2S2 | |
| 5 g | |
| RUDNWZFWWJFUSF-UHFFFAOYSA-M | |
| potassium;(4-methylphenyl)-oxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 23662126 | |
| ≥98.0% (HPLC) | |
| Potassium p-Toluenethiosulfonate |
Safety and Handling
TSCA : Yes