missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxonate 98.0+%, TCI America™

Catalog No. p-7134217
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01645G 5 g
O016425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01645G Supplier TCI America Supplier No. O01645G
Only null left

Chemical Identifiers

CAS 2207-75-2
Molecular Formula C4H2KN3O4
Molecular Weight (g/mol) 195.18
MDL Number MFCD00010565
InChI Key IAPCTXZQXAVYNG-UHFFFAOYSA-M
Synonym potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid
PubChem CID 2723920
ChEBI CHEBI:80230
IUPAC Name potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate
SMILES [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1

Specifications

Melting Point 300°C
Color White
Quantity 5 g
Formula Weight 195.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Oxonate
RTECSNumber RR4580000
TSCA No