missing translation for 'onlineSavingsMsg'
Learn More

Potassium oxalate monohydrate, Honeywell™

Catalog No. p-7072231
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002777 250 g
6002776 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002777 Supplier Honeywell Chemicals Supplier No. 60425250G
Only null left

Purum p.a.,≥99.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Quantity 250 g
Linear Formula (COOK)2 · H2O
UN Number NONH for all modes of transport
Beilstein 3752576
Formula Weight 184.23g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Potassium oxalate monohydrate
missing translation for 'healthHazard2' P280-P301 + P312 + P330