missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate, Monohydrate, Crystal, ACS, 98.5-101.0%, Spectrum™ Chemical

Catalog No. p-7166075
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18612815 125 g Poly Bottle
18612816 500 g Poly Bottle
18612817 2.5 kg Poly Bottle
18612818 12 kg Poly Pail
18612819 50 kg Fiber Drum
5 options

Catalog No. 18612815

Supplier: Spectrum Chemical Mfg Cor P1355125GM

Only null left

Chemical Identifiers

CAS 6487-48-5

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98.5 to 101.0%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.