missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate, Monohydrate, Crystal, ACS, 98.5-101.0%, Spectrum™ Chemical

Catalog No. 18612815
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612815 125 g Poly Bottle
18612816 500 g Poly Bottle
18612817 2.5 kg Poly Bottle
18612818 12 kg Poly Pail
18612819 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612815 Supplier Spectrum Chemical Mfg Cor Supplier No. P1355125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 6487-48-5

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98.5 to 101.0%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.