missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate Monohydrate, ACS Reagent, Reagents

Catalog No. p-200021028
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337818 100 g
01337817 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337818 Supplier Labchem Inc Supplier No. C1287500100C2
Only null left

Potassium Oxalate Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym Dipotassium Ethanedioic Acid
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Boiling Point 365°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.13
Chemical Name or Material Potassium Oxalate Monohydrate, ACS Reagent