missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nonafluoro-1-butanesulfonate 98.0+%, TCI America™
Supplier: TCI America N071125G
Specifications
| Potassium Nonafluoro-1-butanesulfonate | |
| White | |
| MFCD00007515 | |
| Nonafluoro-1-butanesulfonic Acid Potassium Salt, Perfluoro-1-butanesulfonic Acid Potassium Salt, Potassium Perfluoro-1-butanesulfonate | |
| [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
| 338.18 | |
| 338.18 | |
| Crystalline Powder |
| 29420-49-3 | |
| C4F9KO3S | |
| 25 g | |
| LVTHXRLARFLXNR-UHFFFAOYSA-M | |
| potassium nonafluorobutane-1-sulfonate | |
| 23675274 | |
| ≥98.0% (T) |
Chemical Identifiers
| 29420-49-3 | |
| 338.18 | |
| LVTHXRLARFLXNR-UHFFFAOYSA-M | |
| 23675274 | |
| [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| C4F9KO3S | |
| MFCD00007515 | |
| Nonafluoro-1-butanesulfonic Acid Potassium Salt, Perfluoro-1-butanesulfonic Acid Potassium Salt, Potassium Perfluoro-1-butanesulfonate | |
| potassium nonafluorobutane-1-sulfonate |
Safety and Handling
EINECSNumber : (2)-2810
TSCA : Yes