CAS RN 29420-49-3
CAS RN 29420-49-3
IUPAC Name:
potassium nonafluorobutane-1-sulfonate
Synonyms:
potassium nonafluorobutane-1-sulfonate
Molecular Weight (g/mol):
338.18
Molecular Formula:
C4F9KO3S
InChi Key:
LVTHXRLARFLXNR-UHFFFAOYSA-M
SMILES:
[K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
2
of
2
results
1
Verispecâ„¢ PFAS Standards, Certified Reference Material
Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.
2
Potassium Nonafluoro-1-butanesulfonate 98.0+%, TCI Americaâ„¢
CAS: 29420-49-3 Molecular Formula: C4F9KO3S Molecular Weight (g/mol): 338.18 MDL Number: MFCD00007515 InChI Key: LVTHXRLARFLXNR-UHFFFAOYSA-M Synonym: Nonafluoro-1-butanesulfonic Acid Potassium Salt, Perfluoro-1-butanesulfonic Acid Potassium Salt, Potassium Perfluoro-1-butanesulfonate PubChem CID: 23675274 IUPAC Name: potassium nonafluorobutane-1-sulfonate SMILES: [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F