missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, 1 M, Filling Solution, Fisherbrand
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 mL |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Potassium Nitrate, 1M, Fill Solution |