missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Monomethyl Terephthalate 95.0+%, TCI America™
$44.28 - $209.84
Chemical Identifiers
| CAS | 42967-55-5 |
|---|---|
| Molecular Formula | C9H7KO4 |
| Molecular Weight (g/mol) | 218.249 |
| MDL Number | MFCD00058989 |
| InChI Key | DWNDFZTVSFSFTB-UHFFFAOYSA-M |
| Synonym | Terephthalic Acid Monomethyl Ester Potassium Salt |
| PubChem CID | 23683753 |
| IUPAC Name | potassium;4-methoxycarbonylbenzoate |
| SMILES | COC(=O)C1=CC=C(C=C1)C(=O)[O-].[K+] |
Chemical Identifiers
| 42967-55-5 | |
| 218.249 | |
| DWNDFZTVSFSFTB-UHFFFAOYSA-M | |
| 23683753 | |
| COC(=O)C1=CC=C(C=C1)C(=O)[O-].[K+] |
| C9H7KO4 | |
| MFCD00058989 | |
| Terephthalic Acid Monomethyl Ester Potassium Salt | |
| potassium;4-methoxycarbonylbenzoate |
Specifications
| 42967-55-5 | |
| C9H7KO4 | |
| 25 g | |
| DWNDFZTVSFSFTB-UHFFFAOYSA-M | |
| potassium;4-methoxycarbonylbenzoate | |
| 23683753 | |
| ≥95.0% (HPLC,T) | |
| Potassium Monomethyl Terephthalate |
| White | |
| MFCD00058989 | |
| Terephthalic Acid Monomethyl Ester Potassium Salt | |
| COC(=O)C1=CC=C(C=C1)C(=O)[O-].[K+] | |
| 218.249 | |
| 218.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes