missing translation for 'onlineSavingsMsg'
Learn More

Potassium Monomethyl Terephthalate 95.0+%, TCI America™

Catalog No. T108325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T108325G 25 g
T1083500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T108325G Supplier TCI America Supplier No. T108325G
Only null left

Chemical Identifiers

CAS 42967-55-5
Molecular Formula C9H7KO4
Molecular Weight (g/mol) 218.249
MDL Number MFCD00058989
InChI Key DWNDFZTVSFSFTB-UHFFFAOYSA-M
Synonym Terephthalic Acid Monomethyl Ester Potassium Salt
PubChem CID 23683753
IUPAC Name potassium;4-methoxycarbonylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)[O-].[K+]

Specifications

Color White
Quantity 25 g
Formula Weight 218.25
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Monomethyl Terephthalate