missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals

$409.26 - $409.26

Chemical Identifiers

CAS 865-33-8
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.132
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC240605000
SDS View Documents
Thermo Scientific Chemicals
240605000
500 mL Glass bottle
Each for $409.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

865-33-8
70.132
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
potassium;methanolate
CH3KO
BDAWXSQJJCIFIK-UHFFFAOYSA-N
23664618
C[O-].[K+]

Specifications

Colorless
CH3KO
0.1N, exact strength on the certificate of analysis
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
BDAWXSQJJCIFIK-UHFFFAOYSA-N
potassium;methanolate
23664618
Pure
0.85
7°C
Potassium methoxide
865-33-8,67-56-1,108-88-3
CH3OK
0.8500g/mL
Solubility in water: may decompose.
C[O-].[K+]
70.132
70.13
Glass bottle
500 mL
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only