missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite, Crystal, NF, 51.8-57.6%, Spectrum™ Chemical

Catalog No. p-7157953
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605645 500 g Poly Bottle
18605646 2.5 kg Poly Pail
18605648 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605645 Supplier Spectrum Chemical Mfg Cor Supplier No. P1346500GM
Only null left

P1346, 16731-55-8, K2S2O5

Spectrum™ Chemical Potassium Metabisulfite, Crystal, NF is used primarily as an antioxidant in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 51.8 to 57.6%
Grade NF
Packaging Poly Bottle