missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Metabisulfite, Crystal, FCC, 90%, Spectrum™ Chemical
PO191, 16731-55-8, K2S2O5
Supplier: Spectrum Chemical Mfg Cor PO19112KG
Description
Spectrum™ Chemical Potassium Metabisulfite, Crystal, FCC is used as a food preservative and an antioxidant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 16731-55-8 | |
| 12 kg | |
| MFCD00167605 | |
| [K+].[K+].[O-]S(=O)S([O-])(=O)=O | |
| 222.31 | |
| FCC |
| 1 | |
| K2O5S2 | |
| RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
| dipotassium sulfinatosulfonate | |
| 90% | |
| Poly Pail |
Chemical Identifiers
| 16731-55-8 | |
| 222.31 | |
| RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
| [K+].[K+].[O-]S(=O)S([O-])(=O)=O |
| K2O5S2 | |
| MFCD00167605 | |
| dipotassium sulfinatosulfonate |