missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite, Crystal, FCC, 90%, Spectrum™ Chemical

Catalog No. 18605642
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605642 2.5 kg Poly Pail
18605643 12 kg Poly Pail
18605644 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605642 Supplier Spectrum Chemical Mfg Cor Supplier No. PO1912.5KG
Only null left

PO191, 16731-55-8, K2S2O5

Spectrum™ Chemical Potassium Metabisulfite, Crystal, FCC is used as a food preservative and an antioxidant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 90%
Grade FCC
Packaging Poly Pail