missing translation for 'onlineSavingsMsg'
Learn More

Potassium metabisulfite, 95+%

CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol

Supplier:  Thermo Scientific Chemicals 04390836

 View more versions of this product

Catalog No. AA4390836


Only null left
Explore available promotions

Description

Description

It serves as a component of certain photographic developers and its solution is used in photographic fixing, in the textile industry for dyeing and cotton printing. It is utilized to precipitate gold from solution in aqua regia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium metabisulfite
White
Powder
>99%
MFCD00167605
Soluble in water. Insoluble in ethanol.
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
222.31
222.32
Air Sensitive
190°C (decomposition)
16731-55-8
500 g
K2O5S2
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
RWPGFSMJFRPDDP-UHFFFAOYSA-L
dipotassium sulfinatosulfonate
28019
Pungent

Chemical Identifiers

16731-55-8
222.31
RWPGFSMJFRPDDP-UHFFFAOYSA-L
28019
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
K2O5S2
MFCD00167605
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
dipotassium sulfinatosulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

EINECSNumber : 240-795-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only