missing translation for 'onlineSavingsMsg'
Learn More

Potassium L-tartrate hemihydrate, 99%

CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol

Supplier:  Thermo Scientific Chemicals A145560B

 View more versions of this product

Catalog No. AAA145560B


Only null left
Explore available promotions

Description

Description

Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

Potassium L-tartrate hemihydrate
155°C (in -H2O)
200°C to 220°C
MFCD00150136,MFCD00150136
6119985
potassium tartrate dibasic hemihydrate
DFVWEDQSAWCYPI-UHFFFAOYNA-N
(2R,3R)-2,3-dihydroxybutanedioic acid;potassium;hydrate
131855961
99%
6100-19-2
1.98
C8H14K4O13
1000 g
14,7677
Soluble in water.
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O
474.58
235.29 (226.27 Anhydrous)

Chemical Identifiers

6100-19-2
474.58
DFVWEDQSAWCYPI-UHFFFAOYNA-N
131855961
C8H14K4O13
MFCD00150136,MFCD00150136
potassium tartrate dibasic hemihydrate
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-067-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only