missing translation for 'onlineSavingsMsg'
Learn More

Potassium L-tartrate hemihydrate, 99%

Catalog No. p-7021991
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1455630 250 g
AAA145560B 1000 g
AAA145560I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1455630 Supplier Thermo Scientific Chemicals Supplier No. A1455630
Only null left

CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol

Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6100-19-2
Molecular Formula C8H14K4O13
Molecular Weight (g/mol) 474.58
MDL Number MFCD00150136,MFCD00150136
InChI Key DFVWEDQSAWCYPI-UHFFFAOYNA-N
Synonym potassium tartrate dibasic hemihydrate
PubChem CID 131855961
SMILES O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 155°C (in -H2O)
Density 1.98
Boiling Point 200°C to 220°C
Quantity 250 g
Beilstein 6119985
Merck Index 14,7677
Solubility Information Soluble in water.
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;potassium;hydrate
Formula Weight 235.29 (226.27 Anhydrous)
Percent Purity 99%
Chemical Name or Material Potassium L-tartrate hemihydrate
Show More Show Less
EINECSNumber 213-067-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only