missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium L-Aspartate Hydrate 97.0+%, TCI America™
$69.31 - $423.54
Chemical Identifiers
| CAS | 1115-63-5 |
|---|---|
| Molecular Formula | C4H6KNO4 |
| Molecular Weight (g/mol) | 171.19 |
| MDL Number | MFCD00021083 |
| InChI Key | TXXVQZSTAVIHFD-DKWTVANSSA-M |
| Synonym | c4h6no4.k.2h2o, l-aspartic acid potassium salt, dihydrate, potassium 3s-3-amino-3-carboxypropanoate dihydrate |
| PubChem CID | 91886500 |
| IUPAC Name | potassium (2S)-2-amino-3-carboxypropanoate |
| SMILES | [K+].N[C@@H](CC(O)=O)C([O-])=O |
Chemical Identifiers
| 1115-63-5 | |
| 171.19 | |
| TXXVQZSTAVIHFD-DKWTVANSSA-M | |
| 91886500 | |
| [K+].N[C@@H](CC(O)=O)C([O-])=O |
| C4H6KNO4 | |
| MFCD00021083 | |
| c4h6no4.k.2h2o, l-aspartic acid potassium salt, dihydrate, potassium 3s-3-amino-3-carboxypropanoate dihydrate | |
| potassium (2S)-2-amino-3-carboxypropanoate |
Specifications
| 1115-63-5 | |
| C4H6KNO4 | |
| c4h6no4.k.2h2o, l-aspartic acid potassium salt, dihydrate, potassium 3s-3-amino-3-carboxypropanoate dihydrate | |
| [K+].N[C@@H](CC(O)=O)C([O-])=O | |
| 25 g | |
| 91886500 | |
| ≥97.0% (T) | |
| Potassium L-Aspartate Hydrate |
| White | |
| MFCD00021083 | |
| TXXVQZSTAVIHFD-DKWTVANSSA-M | |
| potassium (2S)-2-amino-3-carboxypropanoate | |
| 171.19 | |
| 171.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1548
TSCA : Yes