accessibility menu, dialog, popup

UserName

KNH2(S)HOOOO

CAS RN 1115-63-5

IUPAC Name: potassium (2S)-2-amino-3-carboxypropanoate
Synonyms: potassium aspartate
Molecular Weight (g/mol): 171.19
Molecular Formula: C4H6KNO4
InChi Key: TXXVQZSTAVIHFD-DKWTVANSSA-M
SMILES: [K+].N[C@@H](CC(O)=O)C([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)