missing translation for 'onlineSavingsMsg'
Learn More

Potassium L-Aspartate Hydrate 97.0+%, TCI America™

Catalog No. A092225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A092225G 25 g
A0922500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A092225G Supplier TCI America Supplier No. A092225G

Chemical Identifiers

CAS 1115-63-5
Molecular Formula C4H6KNO4
Molecular Weight (g/mol) 171.19
MDL Number MFCD00021083
InChI Key TXXVQZSTAVIHFD-DKWTVANSSA-M
Synonym c4h6no4.k.2h2o, l-aspartic acid potassium salt, dihydrate, potassium 3s-3-amino-3-carboxypropanoate dihydrate
PubChem CID 91886500
IUPAC Name potassium (2S)-2-amino-3-carboxypropanoate
SMILES [K+].N[C@@H](CC(O)=O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 171.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium L-Aspartate Hydrate
EINECSNumber (9)-1548
TSCA Yes