missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide (White, Free-Flowing Granules), Fisher BioReagents Edge
Edge

Manufacturer:  Fisher BioReagents FLBP367500

Catalog No. BP367-500



Edge

Description

Description

This potassium salt is used in buffers for molecular biology and cell culture applications.
Specifications

Specifications

Potassium Iodide
Vapor Pressure: 1mmHg at 745°C
IK
asmofug e, kali iodide, kisol, knollide, pima, potassium diiodide, potassium iodide, potassium iodide ki, potassium monoiodide, thyro-block
[K+].[I-]
0.002% max.
4875
166
≥99.0%
0.01% max.
Pass Test
3ppm max.
0.2% max.
Poly Bottle
0.005% max.
1mmHg at 745°C
White
6 to 8
7681-11-0
≥99.0 %
15, 7764
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
166.003
CHEBI:8346
Solid
0.002% max.
5ppm max.
0.005% max.
3ppm max.
0.001% max.
0.001% max.
0.005% max.
1330°C
680°C
500g

Chemical Identifiers

7681-11-0
166.003
asmofug e, kali iodide, kisol, knollide, pima, potassium diiodide, potassium iodide, potassium iodide ki, potassium monoiodide, thyro-block
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0

Recommended Storage : RT

SDS Documents
Promotions

Promotions