missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, USP, 99-101.5%, Spectrum™ Chemical

PO185, 7681-11-0, KI

Supplier:  Spectrum Chemical Mfg Cor PO18525KGPLCS4

 View more versions of this product

Catalog No. 18605639

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Iodide, USP is used to treat hyperthyroidism in radiation emergencies and to protect the thyroid gland when certain types of radiopharmaceuticals are used. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7681-11-0
4 x 2.5 kg
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
99 to 101.5%
Poly Bottle
1
IK
[K+].[I-]
166.00
0.01

Chemical Identifiers

7681-11-0
166.00
potassium iodide
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
[K+].[I-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions