missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, USP, 99-101.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Iodide, USP is used to treat hyperthyroidism in radiation emergencies and to protect the thyroid gland when certain types of radiopharmaceuticals are used. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Percent Purity | 99 to 101.5% |
| Grade | USP |
| Loss on Drying | 0.01 |
| Packaging | Amber Glass Bottle |